missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 4.00 N, Ricca Chemical

Catalog No. p-2917120
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
746032 1 L Natural Poly Bottle
74601 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 746032 Supplier Ricca Chemical Company Supplier No. 746032
Only null left

Chemical Identifiers

CAS 1310-73-2

Specifications

Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 4.00N
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hydroxide