missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 25% (w/v) Solution, Spectrum™ Chemical

$105.76 - $1457.74

Chemical Identifiers

CAS 1310-73-2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613661
View Documents
Spectrum Chemical Mfg Cor
S237500ML
500 mL Poly Bottle
Each for $105.76
Only null left
 
18613593
View Documents
Spectrum Chemical Mfg Cor
S2374LTPL
4 L Poly Bottle
Each for $327.47
Only null left
 
18613594
View Documents
Spectrum Chemical Mfg Cor
S23720LT
20 L Cubitainer
Each for $1,457.74
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1310-73-2

Specifications

1310-73-2,7732-18-5
25% ±0.5%
Poly Bottle
500 mL
Chemical Solution
Sodium Hydroxide Solution
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.