missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 10% (w/v) (100 g/L), Ricca Chemical
$64.34 - $554.00
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 39.997 |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| IUPAC Name | sodium;hydroxide |
| SMILES | [OH-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
7260-4
|
Ricca Chemical Company
RI72604 |
120 mL | Natural Poly Bottle |
N/A
|
|
||||
|
7260-16
|
Ricca Chemical Company
726016 |
500 mL | Natural Poly Bottle |
N/A
|
|
||||
|
7260-32
|
Ricca Chemical Company
726032 |
1 L | Natural Poly Bottle |
N/A
|
|
||||
|
7260-1
|
Ricca Chemical Company
72601 |
4 L | Natural Poly Bottle |
N/A
|
|
||||
|
72602-5
|
Ricca Chemical Company
RI726025 |
10 L | Cubitainer |
N/A
|
|
||||
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
Specifications
| 1310-73-2,7732-18-5 | |
| 93.0,9.28 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 9.96 to 10.04% | |
| 10% (w/v) Aqueous Solution (100g/L) | |
| Sodium Hydroxide |
| 89.0,8.91 | |
| HNaO | |
| 120 mL | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| Laboratory | |
| Natural Poly Bottle |