missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 10.0 N, from NF/USP material, Ricca Chemical

Catalog No. R74700501A
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R74700501A 1 L
R74700504A 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R74700501A Supplier Ricca Chemical Company Supplier No. R74700501A
Only null left

Chemical Identifiers

CAS 1310-73-2

Specifications

Color Colorless
CAS Max % 69.32, 30.68
Molecular Formula H2O
Concentration 9.95 to 10.05 N
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Quantity 1 L
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Grade Laboratory
Packaging Natural Poly Bottle
Physical Form Liquid
Assay Normal at 20°C
Chemical Name or Material Sodium Hydroxide Solution
Show More Show Less
ShelfLife 24 months