missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 1.00 N, from NF/USP/EP material, Ricca Chemical
Supplier: Ricca Chemical Company R74500101A
Description
- Tested in an ISO 17025 Accredited Facility
- Test Reports provided with Certified Values and Measurement Uncertainty
- Tight release specifications for low lot-to-lot variability
Spécifications
Sodium Hydroxide Solution | |
1310-73-2,1310-73-2 | |
Normal at 20°C | |
0.999 to 1.001 N | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Laboratory | |
Liquid |
Colorless | |
96.09,3.91 | |
H2O | |
1 L | |
O | |
18.02 g/mol | |
Natural Poly Bottle |
Identifiants chimiques
1310-73-2 | |
18.02 g/mol | |
water |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
O |
Sécurité et manipulation
missing translation for 'shelfLife' : 24 months