missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.1562 N, Ricca Chemical

Catalog No. 735732
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
735732 1 L Natural Poly Bottle
73571 4 L Natural Poly Bottle
73575 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 735732 Supplier Ricca Chemical Company Supplier No. 735732
Only null left
  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 1310-73-2
Molecular Weight (g/mol) 40 g/mol

Specifications

Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.1562N
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hydroxide