missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.125 N (N/8), Ricca Chemical

Catalog No. 73561
Change view
Click to view available options
Quantity:
4 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
73561 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 73561 Supplier Ricca Chemical Company Supplier No. 73561
Only null left
  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 1310-73-2
Molecular Weight (g/mol) 40 g/mol

Specifications

Quantity 4 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.125N (N/8)
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hydroxide