missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.100 N (N/10) in Methanol, Ricca Chemical

Catalog No. p-3252332
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
747232 1 L Natural Poly Bottle
74721 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 747232 Supplier Ricca Chemical Company Supplier No. 747232
Only null left

Chemical Identifiers

CAS 67-56-1 , 1310-73-2
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145

Specifications

Color Colorless
CAS Min % 97.0, 0.53
CAS Max % 100.0, 0.55
Molecular Formula HNaO
Concentration or Composition Notes 0.100Normal
Quantity 1 L
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Methanol
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Show More Show Less