missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrosulfite, 85%, Spectrum™ Chemical

$379.97 - $5893.97

Chemical Identifiers

CAS 7775-14-6
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613514
View Documents
Spectrum Chemical Mfg Cor
S1809500GM
500 g Amber Glass Bottle
Each for $379.97
Only null left
 
18613512
View Documents
Spectrum Chemical Mfg Cor
S180925KG
2.5 kg Amber Glass Bottle
Each for $1,151.35
Only null left
 
18613513
View Documents
Spectrum Chemical Mfg Cor
S180945KGBL
45 kg Fiber Drum
Each for $5,893.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7775-14-6

Specifications

7775-14-6
500 g
Reagent
1
85%
Amber Glass Bottle
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.