missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrosulfite, 85%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S180945KGBL

 View more versions of this product

Catalog No. 18613513


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7775-14-6
45 kg
Reagent
1
85%
Fiber Drum

Chemical Identifiers

7775-14-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.