missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen sulfate monohydrate, Reagent Grade

Catalog No. p-4824952
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1379936 500 g
AA13799A1 1 kg
AA13799A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1379936 Supplier Thermo Scientific Chemicals Supplier No. 01379936
Only null left

CAS: 10034-88-5 | H3NaO5S | 138.07 g/mol

Sodium hydrogen sulfate monohydrate is used in metal finishing and cleaning products. It is used to lower the pH of water for an effective chlorination in swimming pools. It is also used as a urine acidifier to reduce urinary stones in cats. Further, it is used in controlling outbreaks of crown-of-thorns starfish. It is a primary ingredient used in jewelry making to clean the surfaces. In addition to this, it is used as a food additive and serves as a reagent for the decomposition of basic metal oxides by fusion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10034-88-5
Molecular Formula H3NaO5S
Molecular Weight (g/mol) 138.07
MDL Number MFCD00149210
InChI Key JXHZRQHZVYDRGX-UHFFFAOYSA-M
Synonym sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
PubChem CID 23673662
SMILES O.[Na+].OS([O-])(=O)=O

Specifications

Melting Point ∽183°C (decomposition)
Color White
Physical Form Crystals
Quantity 500 g
Solubility Information Soluble in water. Insoluble in ammonia.
IUPAC Name sodium;hydrogen sulfate;hydrate
Formula Weight 138.07 (120.06anhydrous)
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Chemical Name or Material Sodium hydrogen sulfate monohydrate
Show More Show Less
Hazard Category H318
Precautionary Statement P261-P271-P280-P305+P351+P338-P310-P501c
DOTInformation Hazard Class: 8, Packing Group: III
EINECSNumber 231-665-7
RTECSNumber VZ1860000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only