accessibility menu, dialog, popup

UserName

H2ONaOHSOOO

CAS RN 10034-88-5

IUPAC Name: sodium hydrate hydrogen sulfate
Synonyms: sodium bisulfate monohydrate
Molecular Weight (g/mol): 138.07
Molecular Formula: H3NaO5S
InChi Key: JXHZRQHZVYDRGX-UHFFFAOYSA-M
SMILES: O.[Na+].OS([O-])(=O)=O
Grade 
  • (1)
  • (2)
  • (2)
  • (3)
  • (8)
Molecular Weight (g/mol) 
  • (15)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (5)
Physical Form 
  • (5)
  • (5)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (5)
  • (2)
  • (5)
  • (9)
  • (1)
  • (2)
  • (2)
  • (3)
  • (8)
  • (15)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (5)
  • (5)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)