missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrogen Fumarate 98.0+%, TCI America™

Catalog No. F0071500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0071500G 500 g
F007125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0071500G Supplier TCI America Supplier No. F0071500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5873-57-4
Molecular Formula C4H2NaO4
Molecular Weight (g/mol) 137.05
MDL Number MFCD00039097
InChI Key VRVKOZSIJXBAJG-TYYBGVCCSA-L
Synonym sodium hydrogen fumarate, monosodium fumarate, sodium fumarate, maleic acid, sodium salt, maleinian sodu polish, fumaric acid monosodium salt, ccris 7315, 2-butenedioic acid 2z-, sodium salt, sodium e-3-carboxyacrylate, maleinian sodu
PubChem CID 6076814
IUPAC Name sodium (2E)-but-2-enedioate
SMILES [Na+].[O-]C(=O)\C=C\C([O-])=O

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 138.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Hydrogen Fumarate
EINECSNumber (2)-1094
TSCA No