accessibility menu, dialog, popup

UserName

NaOOOO(E)

CAS RN 5873-57-4

IUPAC Name: sodium (2E)-but-2-enedioate
Synonyms: sodium fumarate
Molecular Weight (g/mol): 137.05
Molecular Formula: C4H2NaO4
InChi Key: VRVKOZSIJXBAJG-TYYBGVCCSA-L
SMILES: [Na+].[O-]C(=O)\C=C\C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)