Learn More
Sodium hydrogen fumarate, 98%
CAS: 5873-57-4 | C4H2NaO4 | 137.05 g/mol
$75.89 - $238.51
Chemical Identifiers
| CAS | 5873-57-4 |
|---|---|
| Molecular Formula | C4H2NaO4 |
| Molecular Weight (g/mol) | 137.05 |
| MDL Number | MFCD00039097 |
| InChI Key | VRVKOZSIJXBAJG-TYYBGVCCSA-L |
| Synonym | sodium hydrogen fumarate, monosodium fumarate, sodium fumarate, maleic acid, sodium salt, maleinian sodu polish, fumaric acid monosodium salt, ccris 7315, 2-butenedioic acid 2z-, sodium salt, sodium e-3-carboxyacrylate, maleinian sodu |
| PubChem CID | 6076814 |
| IUPAC Name | sodium;(E)-4-hydroxy-4-oxobut-2-enoate |
| SMILES | [Na+].[O-]C(=O)\C=C\C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAB2468318
|
N/A | 50 g |
Each for $75.89
|
|
|||||
|
AAB2468330
|
N/A | 250 g |
Each for $238.51
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C4H2NaO4 | |
| MFCD00039097 | |
| sodium hydrogen fumarate, monosodium fumarate, sodium fumarate, maleic acid, sodium salt, maleinian sodu polish, fumaric acid monosodium salt, ccris 7315, 2-butenedioic acid 2z-, sodium salt, sodium e-3-carboxyacrylate, maleinian sodu | |
| sodium;(E)-4-hydroxy-4-oxobut-2-enoate |
Specifications
| 5873-57-4 | |
| MFCD00039097 | |
| VRVKOZSIJXBAJG-TYYBGVCCSA-L | |
| sodium;(E)-4-hydroxy-4-oxobut-2-enoate | |
| 6076814 | |
| 98% |
| C4H2NaO4 | |
| sodium hydrogen fumarate, monosodium fumarate, sodium fumarate, maleic acid, sodium salt, maleinian sodu polish, fumaric acid monosodium salt, ccris 7315, 2-butenedioic acid 2z-, sodium salt, sodium e-3-carboxyacrylate, maleinian sodu | |
| [Na+].[O-]C(=O)\C=C\C([O-])=O | |
| 137.05 | |
| 138.06 | |
| Sodium hydrogen fumarate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 227-535-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only