missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen DL-malate, 98%

Catalog No. AAA1942830
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1942830 250 g
AAA1942818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1942830 Supplier Thermo Scientific Chemicals Supplier No. A1942830
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 57467-17-1 | C4H5NaO5 | 156.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57467-17-1
Molecular Formula C4H5NaO5
Molecular Weight (g/mol) 156.07
MDL Number MFCD00798569
InChI Key DOJOZCIMYABYPO-UHFFFAOYNA-M
Synonym sodium hydrogen malate, monosodium malate, sodium malate, l-malic acid, --sodium hydrogen malate, l--malic acid disodium salt, sodium 3-carboxy-3-hydroxypropanoate, 2-hydroxysuccinic acid 1-sodium salt, sodium 3,4-dihydroxy-4-oxo-butanoate, 2-hydroxybutanedioic acid 4-sodium salt
PubChem CID 23678821
IUPAC Name sodium;3,4-dihydroxy-4-oxobutanoate
SMILES [Na+].OC(CC([O-])=O)C(O)=O

Specifications

Melting Point 210°C (decomposition)
Quantity 250 g
Formula Weight 156.07
Percent Purity 98%
Chemical Name or Material Sodium hydrogen DL-malate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only