CAS RN 57467-17-1
IUPAC Name:
sodium 3-carboxy-3-hydroxypropanoate
Synonyms:
sodium malate
Molecular Weight (g/mol):
156.07
Molecular Formula:
C4H5NaO5
InChi Key:
DOJOZCIMYABYPO-UHFFFAOYNA-M
SMILES:
[Na+].OC(CC([O-])=O)C(O)=O
1
–
1
of
1
results
1
Promotions Available
Sodium hydrogen DL-malate, 98%
Sodium hydrogen DL-malate, 98%
CAS: 57467-17-1 Molecular Formula: C4H5NaO5 Molecular Weight (g/mol): 156.07 MDL Number: MFCD00798569 InChI Key: DOJOZCIMYABYPO-UHFFFAOYNA-M Synonym: sodium hydrogen malate,monosodium malate,sodium malate,l-malic acid,--sodium hydrogen malate,l--malic acid disodium salt,sodium 3-carboxy-3-hydroxypropanoate,2-hydroxysuccinic acid 1-sodium salt,sodium 3,4-dihydroxy-4-oxo-butanoate,2-hydroxybutanedioic acid 4-sodium salt PubChem CID: 23678821 IUPAC Name: sodium;3,4-dihydroxy-4-oxobutanoate SMILES: [Na+].OC(CC([O-])=O)C(O)=O