missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrogen Carbonate, MilliporeSigma

Catalog No. 01213175
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
12 kg
25 kg
50 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01213175 25 kg
01213172 500 g
01213170 1 kg
01213169 5 kg
01213174 12 kg
01213171 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01213175 Supplier Merck Emd Millipore Supplier No. 1063299025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium hydrogen carbonate for analysis EMSURE™ ACS,Reag.Ph Eur

Chemical Identifiers

CAS 144-55-8
Molecular Weight (g/mol) 84.01

Specifications

Physical Form Powder
Quantity 25 kg
Percent Purity Not Measured
Grade ACS
Packaging Plastic Drum
Chemical Name or Material Sodium hydrogen carbonate