Learn More
Sodium hydrogen carbonate, 99%
CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
Supplier: Thermo Scientific Chemicals A170050E
Description
Sodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, baking powder, as a mild abrasive and in cleaning compounds. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, baking powder, as a mild abrasive and in cleaning compounds. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.
Solubility
Soluble in water and methanol. Insoluble in ethanol.
Notes
Incompatible with strong acids and strong oxidizing agents.
Specifications
| Sodium hydrogen carbonate | |
| 8.1 to 8.4 | |
| CHNaO3 | |
| 4153970 | |
| sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| sodium hydrogen carbonate | |
| 516892 | |
| 84.01 | |
| Odorless | |
| 2.159 |
| 144-55-8 | |
| 2500 g | |
| MFCD00003528 | |
| 14,8583 | |
| Soluble in water and methanol. Insoluble in ethanol. | |
| [Na+].OC([O-])=O | |
| 84.01 | |
| CHEBI:32139 | |
| 99% | |
| 1.5 |
Chemical Identifiers
| 144-55-8 | |
| 84.01 | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| 516892 | |
| sodium hydrogen carbonate |
| CHNaO3 | |
| MFCD00003528 | |
| sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
| CHEBI:32139 | |
| [Na+].OC([O-])=O |
Safety and Handling
EINECSNumber : 205-633-8
RTECSNumber : VZ0950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only