missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hexanoate 99.0+%, TCI America™

Catalog No. p-7130350
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H011225G 25 g
H0112500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H011225G Supplier TCI America Supplier No. H011225G
Only null left

Chemical Identifiers

CAS 10051-44-2
Molecular Formula C6H11NaO2
Molecular Weight (g/mol) 138.14
MDL Number MFCD00059056
InChI Key UDWXLZLRRVQONG-UHFFFAOYSA-M
Synonym sodium hexanoate, sodium caproate, hexanoic acid, sodium salt, hexanoic acid sodium salt, sodium capronate, sodium n-caproate, sodium n-hexanoate, unii-e00cg11s66, caproic acid sodium salt, hexanoic acid, sodium salt 1:1
PubChem CID 4087444
IUPAC Name sodium hexanoate
SMILES [Na+].CCCCCC([O-])=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 138.14
Percent Purity ≥99.0% (T)
Chemical Name or Material Sodium Hexanoate
EINECSNumber (2)-0611
TSCA Yes