missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hexanitrocobaltate(III), ACS Reagent, Solstice

Catalog No. 60045428
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045428 100 g Poly Bottle
60045429 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045428 Supplier Solstice Supplier No. 233722100G

ACS Reagent

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 13600-98-1
Molecular Formula CoN6Na3O12
Molecular Weight (g/mol) 403.93
MDL Number MFCD00003512
InChI Key BZFWEBJDBATXME-UHFFFAOYSA-N
IUPAC Name trisodium hexanitrocobalttris(ylium)
SMILES [Na+].[Na+].[Na+].[O-][N+](=O)[Co+3]([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 220°C
Color Golden or Orange-Yellow
Quantity 100 g
Linear Formula Na3Co(NO2)6
UN Number UN1479
Formula Weight 403.94g/mol
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Sodium Hexanitrocobaltate(III)
Hazard Statement P220-P261-P280-P305 + P351 + P338-P342 + P311
Precautionary Statement H272-H315-H317-H319-H334-H335-H351
DOTInformation 5.1/PGII
EINECSNumber 237-077-7
RTECSNumber GF9480000