accessibility menu, dialog, popup

UserName

CoNaNaNaNO2NO2NO2NO2NO2NO2

CAS RN 13600-98-1

IUPAC Name: trisodium hexanitrocobalttris(ylium)
Synonyms: trisodium hexanitrocobalttris(ylium)
Molecular Weight (g/mol): 403.93
Molecular Formula: CoN6Na3O12
InChi Key: BZFWEBJDBATXME-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[Na+].[O-][N+](=O)[Co+3]([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)[N+]([O-])=O
Physical Form 
  • (1)
  • (1)
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
Grade 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)