Learn More
Sodium hexacyanoferrate(II) decahydrate, 99%
CAS: 14434-22-1 | C6H20FeN6Na4O10 | 484.06 g/mol
Supplier: Thermo Scientific Chemicals 03966030
Description
Mainly used as important raw material in pharmaceuticals, pigment, tannage, metallurgy and chemical industries. Used in producing iron blue and potassium ferricyanide, surface corrosion protecting for tannage and metal, carburization treating for carbon steel, removing iron in pharmaceutical producing. Used as oxidant, food additive, explosive and chemical reagent. Also used as additive for preventing agglomeration in melt snow in winter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sodium hexacyanoferrate(II) decahydrate | |
| Yellow | |
| Powder | |
| 99% | |
| MFCD00149274 | |
| Soluble in water. Insoluble in alcohol. | |
| O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N | |
| 484.07 (303.92 Anhydrous) | |
| Light yellow powder |
| 82°C-10H2O | |
| 14434-22-1 | |
| 250 g | |
| C6H20FeN6Na4O10 | |
| Sodium ferrocyanide decahydrate | |
| JFSUDVTVQZUDOP-UHFFFAOYSA-N | |
| 484.06 | |
| Odorless | |
| 1.458 |
Chemical Identifiers
| 14434-22-1 | |
| 484.06 | |
| JFSUDVTVQZUDOP-UHFFFAOYSA-N | |
| O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N |
| C6H20FeN6Na4O10 | |
| MFCD00149274 | |
| Sodium ferrocyanide decahydrate |
Safety and Handling
H402-H412: Harmful to aquatic life. Harmful to aquatic life with long lasting effects.
EINECSNumber : 237-081-9
RTECSNumber : LJ8145000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only