accessibility menu, dialog, popup

UserName

CAS RN 14434-22-1

NaNaCFeCCCCCNNNNNNNaNaH2OH2OH2OH2OH2OH2OH2OH2OH2OH2O

CAS RN 14434-22-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 484.06
Molecular Formula: C6H20FeN6Na4O10
InChi Key: JFSUDVTVQZUDOP-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)