missing translation for 'onlineSavingsMsg'
Learn More

Sodium Heptanoate 97.0+%, TCI America™

Supplier:  TCI America E000925G

Quantity 25 g

Catalog No. E000925G

Only null left

Description

Description

Specifications

Chemical Identifiers

10051-45-3
152.17
NMTDPTPUELYEPL-UHFFFAOYSA-M
23663651
[Na+].CCCCCCC([O-])=O
C7H13NaO2
MFCD00070499
Enanthic Acid Sodium Salt, Sodium Enanthate, Heptanoic Acid Sodium Salt
sodium heptanoate

Specifications

Sodium Heptanoate
White
25 g
Enanthic Acid Sodium Salt, Sodium Enanthate, Heptanoic Acid Sodium Salt
[Na+].CCCCCCC([O-])=O
152.17
152.17
Crystalline Powder
10051-45-3
C7H13NaO2
MFCD00070499
NMTDPTPUELYEPL-UHFFFAOYSA-M
sodium heptanoate
23663651
≥97.0% (T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0611

TSCA : No

SDS
Documents

Documents

Promotions

Promotions