accessibility menu, dialog, popup

UserName

NaH3COO

CAS RN 10051-45-3

IUPAC Name: sodium heptanoate
Synonyms: sodium enanthate
Molecular Weight (g/mol): 152.17
Molecular Formula: C7H13NaO2
InChi Key: NMTDPTPUELYEPL-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCC([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results