missing translation for 'onlineSavingsMsg'
Learn More

Sodium Gluconate 99.0+%, TCI America™

Supplier:  TCI America G0041500G

 View more versions of this product

Catalog No. G0041500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Gluconate
206°C
C6H11NaO7
500 g
UPMFZISCCZSDND-JJKGCWMISA-M
sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
23672301
218.14
Crystalline Powder
527-07-1
White-Yellow
MFCD00064210
sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+]
218.137
CHEBI:84997
≥99.0% (T)

Chemical Identifiers

527-07-1
218.137
UPMFZISCCZSDND-JJKGCWMISA-M
23672301
sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C6H11NaO7
MFCD00064210
sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t
CHEBI:84997
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1410

RTECSNumber : LZ5235000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions