missing translation for 'onlineSavingsMsg'
Learn More

Sodium Gluconate 99.0+%, TCI America™

Catalog No. p-7130112
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
G004125G 25 g
G0041500G 500 g
2 options

Catalog No. G004125G

Supplier: TCI America G004125G

Only null left

Chemical Identifiers

CAS 527-07-1
Molecular Formula C6H11NaO7
Molecular Weight (g/mol) 218.137
MDL Number MFCD00064210
InChI Key UPMFZISCCZSDND-JJKGCWMISA-M
Synonym sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t
PubChem CID 23672301
ChEBI CHEBI:84997
IUPAC Name sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+]

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 25 g
Formula Weight 218.14
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Gluconate
EINECSNumber (2)-1410
RTECSNumber LZ5235000
TSCA Yes