missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Gluconate, FCC, 98-102%, Spectrum™ Chemical
SO168, 527-07-1, C6H11NaO7
Supplier: Spectrum Chemical Mfg Cor SO16812KG
Description
Spectrum™ Chemical Sodium Gluconate, FCC is used as surface cleaning agent for metals and glass bottles. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 527-07-1 | |
| Poly Pail | |
| 12 kg | |
| [Na+].OCC(O)C(O)C(O)C(O)C([O-])=O | |
| 218.14 | |
| FCC |
| 1 | |
| C6H11NaO7 | |
| UPMFZISCCZSDND-UHFFFAOYNA-M | |
| sodium 2,3,4,5,6-pentahydroxyhexanoate | |
| 98 to 102% |
Chemical Identifiers
| 527-07-1 | |
| 218.14 | |
| sodium 2,3,4,5,6-pentahydroxyhexanoate |
| C6H11NaO7 | |
| UPMFZISCCZSDND-UHFFFAOYNA-M | |
| [Na+].OCC(O)C(O)C(O)C(O)C([O-])=O |