missing translation for 'onlineSavingsMsg'
Learn More

Sodium Gluconate, FCC, 98-102%, Spectrum™ Chemical

Catalog No. p-7158217
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
Catalog No. Quantity Packaging
18606508 2.5 kg Poly Pail
18606509 12 kg Poly Pail
18606510 45 kg Fiber Drum
3 options

Catalog No. 18606508

Supplier: Spectrum Chemical Mfg Cor SO1682.5KG

Only null left

SO168, 527-07-1, C6H11NaO7

Spectrum™ Chemical Sodium Gluconate, FCC is used as surface cleaning agent for metals and glass bottles. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 527-07-1
Molecular Formula C6H11NaO7
Molecular Weight (g/mol) 218.14
InChI Key UPMFZISCCZSDND-UHFFFAOYNA-M
IUPAC Name sodium 2,3,4,5,6-pentahydroxyhexanoate
SMILES [Na+].OCC(O)C(O)C(O)C(O)C([O-])=O

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 2.5 kg
Percent Purity 98 to 102%
Grade FCC