Learn More
Sodium glucoheptonate
CAS: 31138-65-5 | C7H13NaO8 | 248.163 g/mol
Supplier: Thermo Scientific Chemicals J6681136
Description
Sodium glucoheptonate is used for cleaning metal, mercerizing, paint stripping, and aluminum etching. It is also used to mercerize cotton, linen, hemp, and other plant-based fibers to increase strength and add luster to their overall appearance of threads and fabrics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium glucoheptonate is used for cleaning metal, mercerizing, paint stripping, and aluminum etching. It is also used to mercerize cotton, linen, hemp, and other plant-based fibers to increase strength and add luster to their overall appearance of threads and fabrics.
Solubility
Freely soluble in water
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| Sodium glucoheptonate | |
| 31138-65-5 | |
| 500 g | |
| MFCD00013425 | |
| Freely soluble in water | |
| C(C(C(C(C(C(C(=O)[O-])O)O)O)O)O)O.[Na+] | |
| 248.163 | |
| 248.16 |
| White | |
| Powder | |
| C7H13NaO8 | |
| sodium glucoheptonate, sodium 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoate, d-sodium glucoheptonate, sodium-alpha-glucoheptonate, 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoic acid sodium salt | |
| FMYOMWCQJXWGEN-AXUKCSBQSA-M | |
| sodium;(2R,3R,4R,5S,6R)-2,3,4,5,6,7-hexahydroxyheptanoate | |
| 49853507 |
Chemical Identifiers
| 31138-65-5 | |
| 248.163 | |
| FMYOMWCQJXWGEN-AXUKCSBQSA-M | |
| 49853507 | |
| C(C(C(C(C(C(C(=O)[O-])O)O)O)O)O)O.[Na+] |
| C7H13NaO8 | |
| MFCD00013425 | |
| sodium glucoheptonate, sodium 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoate, d-sodium glucoheptonate, sodium-alpha-glucoheptonate, 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoic acid sodium salt | |
| sodium;(2R,3R,4R,5S,6R)-2,3,4,5,6,7-hexahydroxyheptanoate |
Safety and Handling
EINECSNumber : 250-480-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only