accessibility menu, dialog, popup

UserName

NaHO(R)OH(R)OH(S)OH(R)OHOHOO

CAS RN 31138-65-5

IUPAC Name: sodium (3R,4S,5R,6R)-2,3,4,5,6,7-hexahydroxyheptanoate
Synonyms: sodium (2xi)-D-gluco-heptonate
Molecular Weight (g/mol): 248.16
Molecular Formula: C7H13NaO8
InChi Key: FMYOMWCQJXWGEN-BMZZJELJSA-M
SMILES: [Na+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)C([O-])=O
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results