missing translation for 'onlineSavingsMsg'
Learn More

Sodium glucoheptonate

Catalog No. AAJ6681122
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6681122 100 g
AAJ6681136 500 g
AAJ66811A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6681122 Supplier Thermo Scientific Chemicals Supplier No. J6681122

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 31138-65-5 ∣ C7H13NaO8 ∣ 248.163 g/mol

Sodium glucoheptonate is used for cleaning metal, mercerizing, paint stripping, and aluminum etching. It is also used to mercerize cotton, linen, hemp, and other plant-based fibers to increase strength and add luster to their overall appearance of threads and fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications

Sodium glucoheptonate is used for cleaning metal, mercerizing, paint stripping, and aluminum etching. It is also used to mercerize cotton, linen, hemp, and other plant-based fibers to increase strength and add luster to their overall appearance of threads and fabrics.

Solubility

Freely soluble in water

Notes

Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 31138-65-5
Molecular Formula C7H13NaO8
Molecular Weight (g/mol) 248.163
MDL Number MFCD00013425
InChI Key FMYOMWCQJXWGEN-AXUKCSBQSA-M
Synonym sodium glucoheptonate, sodium 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoate, d-sodium glucoheptonate, sodium-alpha-glucoheptonate, 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoic acid sodium salt
PubChem CID 49853507
IUPAC Name sodium;(2R,3R,4R,5S,6R)-2,3,4,5,6,7-hexahydroxyheptanoate
SMILES C(C(C(C(C(C(C(=O)[O-])O)O)O)O)O)O.[Na+]

Specifications

Color White
Physical Form Powder
Quantity 100 g
Solubility Information Freely soluble in water
Formula Weight 248.16
Grade Ungraded
Chemical Name or Material Sodium glucoheptonate
EINECSNumber 250-480-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only