missing translation for 'onlineSavingsMsg'
Learn More

Sodium Glucoheptonate Dihydrate 98.0+%, TCI America™

Catalog No. G0214500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0214500G 500 g
G021425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0214500G Supplier TCI America Supplier No. G0214500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31138-65-5
Molecular Formula C7H13NaO8
Molecular Weight (g/mol) 248.163
MDL Number MFCD00013425
InChI Key FMYOMWCQJXWGEN-AXUKCSBQSA-M
Synonym sodium glucoheptonate, sodium 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoate, d-sodium glucoheptonate, sodium-alpha-glucoheptonate, 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoic acid sodium salt
PubChem CID 49853507
IUPAC Name sodium;(2R,3R,4R,5S,6R)-2,3,4,5,6,7-hexahydroxyheptanoate
SMILES C(C(C(C(C(C(C(=O)[O-])O)O)O)O)O)O.[Na+]

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 248.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Glucoheptonate Dihydrate