missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoride, Powder, USP, EP, BP, 98.5-100.5%, Spectrum™ Chemical

SO167, 7681-49-4, NaF

$939.53 - $13893.20

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
IUPAC Name sodium fluoride
SMILES [F-].[Na+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18606503
View Documents
Spectrum Chemical Mfg Cor
SO167500GM
500 g Poly Bottle
Each for $939.53
Only null left
 
18606504
View Documents
Spectrum Chemical Mfg Cor
SO16725KG
2.5 kg Poly Bottle
Each for $3,201.27
Only null left
 
18606505
View Documents
Spectrum Chemical Mfg Cor
SO16712KG
12 kg Poly Pail
Each for $6,531.80
Only null left
 
18606506
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
SO16745KGBL
45 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Sodium Fluoride, Powder, USP, EP, BP is used to prevent cavities making teeth stronger and more resistant to decay caused by acid and bacteria. Toothpaste being the preferred delivery method. All Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

7681-49-4
41.99
PUZPDOWCWNUUKD-UHFFFAOYSA-M
[F-].[Na+]
FNa
MFCD00003524
sodium fluoride

Specifications

7681-49-4
500 g
MFCD00003524
[F-].[Na+]
41.99
Multi-Compendial
Poly Bottle
1
FNa
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride
98.5 to 100.5%
0.005
Clear
Videos
SDS
Documents

Documents