missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoride, Powder, USP, EP, BP, 98.5-100.5%, Spectrum™ Chemical

Catalog No. 18606503
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606503 500 g Poly Bottle
18606504 2.5 kg Poly Bottle
18606505 12 kg Poly Pail
18606506 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606503 Supplier Spectrum Chemical Mfg Cor Supplier No. SO167500GM
Only null left

SO167, 7681-49-4, NaF

Spectrum™ Chemical Sodium Fluoride, Powder, USP, EP, BP is used to prevent cavities making teeth stronger and more resistant to decay caused by acid and bacteria. Toothpaste being the preferred delivery method. All Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 98.5 to 100.5%
Grade Multi-Compendial
Loss on Drying 0.005
Packaging Poly Bottle
Appearance Clear