missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoride, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019697
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019697 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019697 Supplier Merck Emd Millipore Supplier No. PHR14081G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Sodium fluoride is a metallic halide. It has been reported as a dental caries preventive agent. It is the most widely employed chemical preservative for the preservation of glucose in blood.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 7681-49-4
Vapor Pressure 1.4 mmHg (0°C)
Physical Form Neat
Linear Formula NaF
MDL Number MFCD00003524
Melting Point 993°C
Synonym Sodium fluoride
Quantity 1 g
Formula Weight 41.99
Grade Certified Reference Material