missing translation for 'onlineSavingsMsg'
Learn More

Sodium Flavonol-2'-sulfonate Hydrate 98.0+%, TCI America™

Catalog No. F01841G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F01841G Supplier TCI America Supplier No. F01841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77125-87-2
Molecular Formula C15H9NaO6S
Molecular Weight (g/mol) 340.281
MDL Number MFCD00059757
InChI Key BAUFCGCZFJQCFG-UHFFFAOYSA-M
Synonym 3-Hydroxyflavone-2′C-sulfonic Acid Sodium Salt, Flavonol-2′C-sulfonic Acid Sodium Salt
PubChem CID 44630295
IUPAC Name sodium;2-(3-hydroxy-4-oxochromen-2-yl)benzenesulfonate
SMILES C1=CC=C2C(=C1)C(=O)C(=C(O2)C3=CC=CC=C3S(=O)(=O)[O-])O.[Na+]

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 340.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Flavonol-2′-sulfonate Hydrate [for Determination of Sn, Zr]