accessibility menu, dialog, popup

UserName

OOSOOOOHNa

CAS RN 77125-87-2

IUPAC Name: sodium 2-(3-hydroxy-4-oxo-4H-chromen-2-yl)benzene-1-sulfonate
Synonyms: sodium 2-(3-hydroxy-4-oxochromen-2-yl)benzenesulfonate
Molecular Weight (g/mol): 340.28
Molecular Formula: C15H9NaO6S
InChi Key: BAUFCGCZFJQCFG-UHFFFAOYSA-M
SMILES: [Na+].OC1=C(OC2=CC=CC=C2C1=O)C1=CC=CC=C1S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results