missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium ferrocyanide decahydrate
CAS: 14434-22-1 | C6H20FeN6Na4O10 | 484.06 g/mol
$69.35 - $69.35
Chemical Identifiers
| CAS | 14434-22-1 |
|---|---|
| Molecular Formula | C6H20FeN6Na4O10 |
| Molecular Weight (g/mol) | 484.06 |
| MDL Number | MFCD00149274 |
| InChI Key | JFSUDVTVQZUDOP-UHFFFAOYSA-N |
| SMILES | O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC459292500
|
Thermo Scientific Chemicals
459292500 |
250 g |
Each for $69.35
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14434-22-1 | |
| 484.06 | |
| JFSUDVTVQZUDOP-UHFFFAOYSA-N |
| C6H20FeN6Na4O10 | |
| MFCD00149274 | |
| O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N |
Specifications
| 14434-22-1 | |
| C6H20FeN6Na4O10 | |
| Solubility in water: 369g/L (20°C). Other solubilities: insoluble in alcohol | |
| O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N | |
| 484.06 | |
| Sodium ferrocyanide decahydrate |
| 250 g | |
| MFCD00149274 | |
| JFSUDVTVQZUDOP-UHFFFAOYSA-N | |
| 484.06 | |
| 1% max. (free moisture) |
RUO – Research Use Only