missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ferrocyanide Decahydrate, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S127412KG

 View more versions of this product

Catalog No. 18613483


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

14434-22-1
12 kg
Reagent
1
99%
Poly Pail

Chemical Identifiers

14434-22-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.