missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ethylxanthate 97.0+%, TCI America™

Catalog No. E019525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E019525G 25 g
E0195500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E019525G Supplier TCI America Supplier No. E019525G
Only null left

Chemical Identifiers

CAS 140-90-9
Molecular Formula C3H5NaOS2
Molecular Weight (g/mol) 144.182
MDL Number MFCD00068314
InChI Key RZFBEFUNINJXRQ-UHFFFAOYSA-M
Synonym Ethylxanthic Acid Sodium Salt, Sodium O-Ethyl Dithiocarbonate, Sodium Xanthogenate
PubChem CID 23690437
IUPAC Name sodium;ethoxymethanedithioate
SMILES CCOC(=S)[S-].[Na+]

Specifications

Color Yellow
Quantity 25 g
Formula Weight 144.18
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Ethylxanthate
EINECSNumber (2)-1287
TSCA Yes