missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ethyl Sulfate 98.0+%, TCI America™

Catalog No. E027725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E027725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E027725G Supplier TCI America Supplier No. E027725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 546-74-7
Molecular Formula C2H5NaO4S
Molecular Weight (g/mol) 148.108
MDL Number MFCD00067412
InChI Key RRLOOYQHUHGIRJ-UHFFFAOYSA-M
Synonym Ethanesulfuric Acid Sodium Salt, Ethylsulfuric Acid Sodium Salt
PubChem CID 23680278
IUPAC Name sodium;ethyl sulfate
SMILES CCOS(=O)(=O)[O-].[Na+]

Specifications

Color White
pH 5.0 - 8
Quantity 25 g
Formula Weight 148.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Ethyl Sulfate