CAS RN 546-74-7
IUPAC Name:
sodium ethyl sulfate
Synonyms:
sodium ethyl sulfate
Molecular Weight (g/mol):
148.11
Molecular Formula:
C2H5NaO4S
InChi Key:
RRLOOYQHUHGIRJ-UHFFFAOYSA-M
SMILES:
[Na+].CCOS([O-])(=O)=O
1
–
1
of
1
results
1
Sodium Ethyl Sulfate 98.0+%, TCI America™
CAS: 546-74-7 Molecular Formula: C2H5NaO4S Molecular Weight (g/mol): 148.108 MDL Number: MFCD00067412 InChI Key: RRLOOYQHUHGIRJ-UHFFFAOYSA-M Synonym: Ethanesulfuric Acid Sodium Salt, Ethylsulfuric Acid Sodium Salt PubChem CID: 23680278 IUPAC Name: sodium;ethyl sulfate SMILES: CCOS(=O)(=O)[O-].[Na+]