missing translation for 'onlineSavingsMsg'
Learn More

Sodium Erythorbate, FCC, 98-100.5%, Spectrum™ Chemical

Catalog No. p-7158215
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606497 125 g Poly Bottle
18606498 500 g Poly Bottle
18606499 2.5 kg Poly Pail
18606500 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606497 Supplier Spectrum Chemical Mfg Cor Supplier No. S1223125GM
Only null left

S1223, 6381-77-7, C6H7NaO6·H2O

Spectrum™ Chemical Sodium Erythorbate, FCC is used as an antioxidant preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6381-77-7
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
InChI Key QNSCGSPRKIPKTN-RKJRWTFHSA-N
IUPAC Name sodium (2R)-2-[(2R)-4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl]-2-hydroxyethan-1-olate
SMILES [Na+].O[C@H](C[O-])[C@H]1OC(O)=C(O)C1=O

Specifications

CAS Max % 1
Loss on Drying 0.0025
Packaging Poly Bottle
Quantity 125 g
Percent Purity 98 to 100.5%
Grade FCC