missing translation for 'onlineSavingsMsg'
Learn More

Sodium Electrode, ISA, Fisherbrand

Catalog No. ACCU0832500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACCU0832500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACCU0832500 Supplier Ricca Chemical Company Supplier No. ACCU0832500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Sodium Electrode ISA
Packaging Natural Poly Bottle
CAS 12125-02-9, 1336-21-6, 7732-18-5
Odor Characteristic ammonia odor
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
Solubility Miscible with water
Solution Type Calibration Solution
Quantity 500 mL
Grade Laboratory
Show More Show Less