missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate 97.0+%, TCI America™

Catalog No. p-7126714
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
D140325G 25 g
D1403500G 500 g
2 options

Catalog No. D140325G

Supplier: TCI America D140325G

Only null left

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Melting Point 207°C
Color White-Yellow
Quantity 25 g
Formula Weight 288.38
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Dodecyl Sulfate
EINECSNumber (2)-1679
RTECSNumber WT1050000
TSCA Yes