missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate, Molecular Grade, Promega™

Catalog No. PRV6551
Encompass
Change view
Click to view available options
Quantity:
100 g
100 mL
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV6551 100 mL
PRH5113 100 g
PRH5114 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRV6551 Supplier Promega Corporation Supplier No. V6551
Only null left

A detergent to denature proteins.

  • Quality tested and certified free of DNase and RNase activity

Chemical Identifiers

CAS 7732-18-5

Specifications

Quantity 100 mL
Grade Molecular Biology
Chemical Name or Material Sodium Dodecyl Sulfate
Recommended Storage 15 to 30°C