missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dodecyl sulfate, >99%, MP Biomedicals™
Specifications
Specifications
| Color | White |
| Density | >1.1 at 20°C |
| Quantity | 1 kg |
| IUPAC Name | sodium dodecyl sulfate |
| Formula Weight | 288.38 |
| Percent Purity | >99% |
| Physical Form | Granular Powder |
| Chemical Name or Material | Sodium dodecyl sulfate |