missing translation for 'onlineSavingsMsg'
Learn More

Sodium DL-2-Hydroxybutyrate 98.0+%, TCI America™

Catalog No. H03875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03875G 5 g
H038725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03875G Supplier TCI America Supplier No. H03875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5094-24-6
Molecular Formula C4H7NaO3
Molecular Weight (g/mol) 126.09
MDL Number MFCD00004565
InChI Key MOSCXNXKSOHVSQ-UHFFFAOYNA-M
Synonym sodium 2-hydroxybutanoate, sodium 2-hydroxybutyrate, sodium dl-2-hydroxybutyrate, 2-hydroxybutyric acid sodium salt, dl-2-hydroxybutyric acid sodium salt, sodium hydroxybutyrate, acmc-1btqj, sodium, a-hydroxybutyrate, sodium 2-oxidanylbutanoate, sodiumdl-2-hydroxybutyrate
PubChem CID 23663641
IUPAC Name sodium 2-hydroxybutanoate
SMILES [Na+].CCC(O)C([O-])=O

Specifications

Color White
Quantity 5 g
Formula Weight 126.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium DL-2-Hydroxybutyrate